MilliporeSigma
  • Decreased expression of LASS2 is associated with worse prognosis in meningiomas.
Decreased expression of LASS2 is associated with worse prognosis in meningiomas.
Journal of neuro-oncology (2014-04-18)
Rong-Hu Ke, Yin Wang, Yin Mao, Jun Zhang, Ji Xiong
ABSTRACT
Homo sapiens longevity assurance homolog 2 of yeast LAG1 (LASS2) has been indicated to have a critical role in various tumors. In the study, we aimed to evaluate the LASS2 expression level in prognostic significance and compare it with commonly used biomarkers: Ki-67, p53 and progesterone receptor (PR) for patients with meningiomas. Firstly, 50 fresh tissues and 143 paraffin-embedded meningiomas samples were analyzed for LASS2 expression by quantitative PCR and immunohistochemistry (IHC), respectively. Subsequently, LASS2 immunostaining was evaluated for its clinical significance. Furthermore, Correlations of LASS2 expression with common biomarkers were assessed. Both PCR and IHC results showed LASS2 was downregulated in high-grade meningiomas in comparison with that of grade I or normal brain (all P < 0.01). IHC results demonstrated LASS2 intensity distribution (ID) score was significantly correlated with tumor size, brain invasion, tumor recurrence and clinical course (all P < 0.01), whereas no correlation of LASS2 ID score with sex or Simpson grade. Moreover, lower LASS2 ID score was strikingly associated with shorter overall and progression-free survival (P < 0.01). Pearson's analysis revealed the ID score was significantly reversely associated with Ki-67 and p53 but not with PR. More importantly,multivariate analyses revealed that LASS2 was an independent prognostic factor (P < 0.05). To our knowledge, it is the first time to investigate the expression of LASS2 and identify it as a potential biomarker for prognosis in meningiomas.
Sigma-Aldrich
Anti-TP53 (ab3) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Hydrogen peroxide solution, for ultratrace analysis, 30 % (w/w)
Progesterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology