MilliporeSigma
Nrf2 deficiency impairs fracture healing in mice.
Calcified tissue international (2014-08-07)
Sebastian Lippross, Rainer Beckmann, Nadine Streubesand, Ferda Ayub, Mersedeh Tohidnezhad, Graeme Campbell, Yuet Wai Kan, Fischer Horst, Tolga Taha Sönmez, Deike Varoga, Philipp Lichte, Holger Jahr, Thomas Pufe, Christoph Jan Wruck
ABSTRACT
Oxidative stress plays an important role in wound healing but data relating oxidative stress to fracture healing are scarce. Nuclear factor erythroid 2-related factor 2 (Nrf2) is the major transcription factor that controls the cellular defence essential to combat oxidative stress by regulating the expression of antioxidative enzymes. This study examined the impact of Nrf2 on fracture healing using a standard closed femoral shaft fracture model in wild-type (WT) and Nrf2-knockout (Nrf2-KO)-mice. Healing was evaluated by histology, real-time RT-PCR, µCT and biomechanical measurements. We showed that Nrf2 expression is activated during fracture healing. Bone healing and remodelling were retarded in the Nrf2-KO compared to the WT-mice. Nrf2-KO-mice developed significantly less callus tissue compared to WT-mice. In addition, biomechanical testing demonstrated lower strength against shear stress in the Nrf2-KO-group compared to WT. The expression of vascular endothelial growth factor (VEGF) and osteocalcin is reduced during fracture healing in Nrf2-KO-mice. Taken together, our results demonstrate that Nrf2 deficiency in mice results in impaired fracture healing suggesting that Nrf2 plays an essential role in bone regeneration. Pharmacological activation of Nrf2 may have therapeutic potential for the enhancement of fracture healing.
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
Anti-NFE2L2 antibody produced in rabbit
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis