MilliporeSigma
  • NK1 receptor antagonists as a new treatment for corneal neovascularization.
NK1 receptor antagonists as a new treatment for corneal neovascularization.
Investigative ophthalmology & visual science (2014-09-18)
Fabio Bignami, Chiara Giacomini, Anna Lorusso, Andrea Aramini, Paolo Rama, Giulio Ferrari
ABSTRACT
To determine whether the inhibition of Substance P (SP) activity can reduce corneal neovascularization (CNV) by means of local administration of high-affinity, competitive, tachykinin 1 receptor (NK1R) antagonists Lanepitant and Befetupitant. We performed a safety and efficacy study by using (1) two different C57BL/6 mouse models of CNV: alkali burn and sutures; (2) different concentrations; and (3) different routes of administration: topical or subconjunctival. Clinical examination endpoints, SP levels, CNV index, and leukocyte infiltration were measured. Substance P increased after injury in the corneal epithelium of both CNV models, and later in the suture model. Topical Lanepitant was nontoxic to the ocular surface and effective in reducing hemangiogenesis and lymphangiogenesis, corneal SP levels, and leukocyte infiltration, as soon as 4 days later in the alkali burn model. Topical Lanepitant, up to 7 days, was ineffective in the suture model. However, subconjunctival Lanepitant was effective in reducing lymphatic CNV, leukocyte infiltration, and SP levels in the suture model, after 10 days. Additionally, in the alkali burn model, subconjunctival Lanepitant significantly reduced blood CNV, corneal perforation rate, opacity, and leukocyte infiltration, and improved tear secretion. Finally, topical application of Befetupitant reduced CNV in the alkali burn model but was toxic owing to the vehicle (dimethyl sulfoxide [DMSO]); hence, Befetupitant was not tested in the suture model. The NK1R antagonist Lanepitant is safe for the ocular surface and effective in reducing both corneal hemangiogenesis and lymphangiogenesis, and leukocyte infiltration. We suggest that inhibition of NK1R may represent an adjunctive tool in the treatment of CNV.
Sigma-Aldrich
Anti-Substance P Antibody, pain, serum, Chemicon®
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O