MilliporeSigma
  • Simulated microgravity-induced mitochondrial dysfunction in rat cerebral arteries.
Simulated microgravity-induced mitochondrial dysfunction in rat cerebral arteries.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-03-08)
Ran Zhang, Hai-Hong Ran, Li-Li Cai, Li Zhu, Jun-Fang Sun, Liang Peng, Xiao-Juan Liu, Lan-Ning Zhang, Zhou Fang, Yong-Yan Fan, Geng Cui
ABSTRACT
Exposure to microgravity results in cardiovascular deconditioning, and cerebrovascular oxidative stress injury has been suggested to occur. To elucidate the mechanism for this condition, we investigated whether simulated microgravity induces mitochondrial dysfunction in rat arteries. Four-week hindlimb unweighting (HU) was used to simulate microgravity in rats. Mitochondrial reactive oxygen species (ROS), mitochondrial membrane potential (Δψm), mitochondrial permeability transition pore (mPTP) opening, mitochondrial respiratory control ratio (RCR), MnSOD/GPx activity and expression, and mitochondrial malondialdehyde (MDA) were examined in rat cerebral and mesenteric VSMCs. Compared with the control rats, mitochondrial ROS levels, mPTP opening, and MDA content increased significantly (P<0.001, P<0.01, and P<0.01, respectively), Δψm, RCR, MnSOD/GPx activity (P<0.001 for Δψm and RCR; P<0.05 for MnSOD; and P<0.001 for GPx activity) and protein abundance of mitochondrial MnSOD/GPx-1 decreased (P<0.001 for MnSOD and GPx-1) in HU rat cerebral but not mesenteric arteries. Chronic treatment with NADPH oxidase inhibitor apocynin and mitochondria-targeted antioxidant mitoTempol promoted recovery of mitochondrial function in HU rat cerebral arteries, but exerted no effects on HU rat mesenteric arteries. Therefore, simulated microgravity resulted in cerebrovascular mitochondrial dysfunction, and crosstalk between NADPH oxidase and mitochondria participated in the process.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, ~1 M in H2O