MilliporeSigma
  • Reduction of sulfamethoxazole hydroxylamine (SMX-HA) by the mitochondrial amidoxime reducing component (mARC).
Reduction of sulfamethoxazole hydroxylamine (SMX-HA) by the mitochondrial amidoxime reducing component (mARC).
Chemical research in toxicology (2014-08-30)
Gudrun Ott, Birte Plitzko, Carmen Krischkowski, Debora Reichmann, Florian Bittner, Ralf R Mendel, Thomas Kunze, Bernd Clement, Antje Havemeyer
ABSTRACT
Under high dose treatment with sulfamethoxazole (SMX)/trimethoprim (TMP), hypersensitivity reactions occur with a high incidence. The mechanism of this adverse drug reaction is not fully understood. Several steps in the toxification pathway of SMX were investigated. The aim of our study was to investigate the reduction of sulfamethoxazole hydroxylamine (SMX-HA) in this toxification pathway, which can possibly be catalyzed by the mARC-containing N-reductive enzyme system. Western blot analyses of subcellular fractions of porcine tissue were performed with antibodies against mARC-1, mARC-2, cytochrome b5 type B, and NADH cytochrome b5 reductase. Incubations of porcine and human subcellular tissue fractions and of the heterologously expressed human components of the N-reductive enzyme system were carried out with SMX-HA. mARC-1 and mARC-2 knockdown was performed in HEK-293 cells. Kinetic parameters of the heterologously expressed human protein variants V96L, A165T, M187 K, C246S, D247H, and M268I of mARC-1 and G244S and C245W of mARC-2 and N-reductive activity of 2SF, D14G, K16E, and T22A of cytochrome b5 type B were analyzed. Western blot analyses were consistent with the hypothesis that the mARC-containing N-reductive enzyme system might be involved in the reduction of SMX-HA. In agreement with these results, highest reduction rates were found in mitochondrial subcellular fractions of porcine tissue and in the outer membrane vesicle (OMV) of human liver tissue. Knockdown studies in HEK-293 cells demonstrated that mARC-1 and mARC-2 were capable of reducing SMX-HA in cell metabolism. Investigations with the heterologously expressed human mARC-2 protein showed a higher catalytic efficiency toward SMX-HA than mARC-1, but none of the investigated human protein variants showed statistically significant differences of its N-reductive activity and was therefore likely to participate in the pathogenesis of hypersensitivity reaction under treatment with SMX.
Product Description
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source