MilliporeSigma
  • Small intestinal contrast ultrasonography for the detection of small bowel complications in Crohn's disease: correlation with intraoperative findings and magnetic resonance enterography.
Small intestinal contrast ultrasonography for the detection of small bowel complications in Crohn's disease: correlation with intraoperative findings and magnetic resonance enterography.
Journal of gastroenterology and hepatology (2014-08-30)
Shankar Kumar, Aishah Hakim, Christopher Alexakis, Vivek Chhaya, Demitrios Tzias, James Pilcher, Johnny Vlahos, Richard Pollok
ABSTRACT
In evaluating small bowel Crohn's disease (CD), small intestine contrast-enhanced ultrasonography (SICUS) is emerging as an alternative to magnetic resonance enterography (MRE). This retrospective study compared the diagnostic accuracy of SICUS and MRE with surgical findings, and their level of agreement. We identified a cohort of CD patients investigated by either SICUS and/or MRE that subsequently required resective bowel surgery within 6 months. The accuracy and agreement of SICUS and MRE to detect small bowel complications were compared with intraoperative findings using kappa coefficient (κ). Agreement between SICUS and MRE in those undergoing both modalities was also assessed. A total of 67 patients were evaluated; 25 underwent SICUS and 17 underwent MRE prior to surgery. Another 25 patients underwent both SICUS and MRE. When compared with intraoperative findings, the sensitivity of SICUS and MRE was 87.5% and 100%, respectively, in detecting strictures, 87.7% and 66.7% for fistulae, 100% for both in identifying abscesses, 100% and 66.7% for bowel dilatation, and 94.7% and 81.8% in defining bowel wall thickening. When correlating SICUS and MRE with surgery, there was a high level of agreement in localizing strictures (κ = 0.75, 0.88, respectively), fistulae (κ = 0.82, 0.79) and abscesses (κ = 0.87, 0.77). Concordance between SICUS and MRE was substantial or almost complete in identifying stricturing disease (κ = 0.84), their number and location (κ = 0.85), fistulae (κ = 0.65), and mucosal thickening (κ = 0.61). SICUS accurately identified small bowel complications and correlated well with MRE and intraoperative findings. SICUS offers an alternative in the preoperative assessment of CD.
Product Description
Mannitol, European Pharmacopoeia (EP) Reference Standard
Metoclopramide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Scopolamine N-butyl bromide, ≥98% (TLC), powder
Supelco
Metoclopramide, VETRANAL®, analytical standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Hyoscine butylbromide, European Pharmacopoeia (EP) Reference Standard