MilliporeSigma
  • Discrepancies between c-Kit positive and Ano1 positive ICC-SMP in the W/Wv and wild-type mouse colon; relationships with motor patterns and calcium transients.
Discrepancies between c-Kit positive and Ano1 positive ICC-SMP in the W/Wv and wild-type mouse colon; relationships with motor patterns and calcium transients.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2014-07-22)
Xuan-Yu Wang, Ji-Hong Chen, Kongling Li, Yong F Zhu, George W J Wright, Jan D Huizinga
ABSTRACT
Interstitial cells of Cajal associated with the submuscular plexus (ICC-SMP) generate omnipresent slow-wave activity in the colon and are associated with prominent motor patterns. Our aim was to investigate colon motor dysfunction in W/W(v) mice in which the ICC are reportedly reduced. Whole organ colon motility was studied using spatio-temporal mapping; immunohistochemical staining was carried out for c-Kit and Ano1; calcium imaging was applied to ICC-SMP. Discrepancies between Ano1 and c-Kit staining were found in both wild-type and W/W(v) colon. ICC-SMP were reduced to ~50% in the W/W(v) mouse colon according to c-Kit immunohistochemistry, but Ano1 staining indicated a normal network of ICC-SMP. The latter was consistent with rhythmic calcium transients occurring at the submucosal border of the colon in W/W(v) mice, similar to the rhythmic transients in wild-type ICC-SMP. Furthermore, the motor pattern associated with ICC-SMP pacemaking, the so-called 'ripples' were normal in the W/W(v) colon. c-Kit is not a reliable marker for quantifying ICC-SMP in the mouse colon. Ano1 staining revealed a normal network of ICC-SMP consistent with the presence of a normal 'ripples' motor pattern. We detected a class of Ano1 positive c-Kit negative cells that do not depend on Kit expression for maintenance, a feature shared with ICC progenitors.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard