MilliporeSigma
  • Myeloid-derived suppressor cells attenuate TH1 development through IL-6 production to promote tumor progression.
Myeloid-derived suppressor cells attenuate TH1 development through IL-6 production to promote tumor progression.
Cancer immunology research (2014-04-30)
Hirotake Tsukamoto, Ryutaro Nishikata, Satoru Senju, Yasuharu Nishimura
ABSTRACT
Collaborative action between tumor cells and host-derived suppressor cells leads to peripheral tolerance of T cells to tumor antigens. Here, we showed that in tumor-bearing mice, generation of tumor antigen-specific effector T-helper cells (TH1) was significantly attenuated, and impaired TH1 differentiation was restored by the temporal blockade of interleukin (IL)-6 activity at the T-cell priming phase. Furthermore, we found that Gr-1(+) myeloid-derived suppressor cells (MDSC) served as a source of IL-6 in tumor-bearing mice. Adoptive transfer of effector CD4(+) T cells revealed that MDSC-sensitized effector CD4(+) T cells were less potent in mounting antitumor immune responses, although effector T cells generated together with Gr-1(+) cells from tumor-free mice eradicated established tumors. CD8(+) T cells, IFN-γ, and MHC-class II expression in host mice were indispensable for the antitumor activity initiated by effector CD4(+) T cells. Despite comparable suppressive activity of IL-6(+/+) and IL-6(-/-) MDSC on primary T-cell activation, transfer of IL-6(+/+) MDSC, but not IL-6(-/-) MDSC, dampened the efficient induction of effector TH1 cells and counteracted CD4(+) T cell-mediated antitumor immunity including cognate help for CD8(+) T cells in vivo. These findings suggest that, apart from the inhibitory effects on primary T-cell activation, MDSC promote tumor progression by attenuating functional differentiation of tumor-specific CD4(+) T cells into effector TH1 cells through IL-6 production to promote tumor progression. This novel mode of MDSC-induced tolerance of effector CD4(+) T cells should be considered as the basis for the rational design of effective T cell-mediated antitumor therapies.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
NG-Methyl-L-arginine acetate salt, ≥98% (TLC)