MilliporeSigma
  • Cardiotonic steroids induce anti-angiogenic and anti-proliferative profiles in first trimester extravillous cytotrophoblast cells.
Cardiotonic steroids induce anti-angiogenic and anti-proliferative profiles in first trimester extravillous cytotrophoblast cells.
Placenta (2014-08-26)
J C Ehrig, D Horvat, S R Allen, R O Jones, T J Kuehl, M N Uddin
ABSTRACT
Preeclampsia (preE), is characterized by abnormal placental invasion and function. Marinobufagenin (MBG), a cardiotonic steroid (CTS), inhibits cytotrophoblast (CTB) cell functions that are critical for normal placental development. This study tests the hypothesis that CTSs induce anti-angiogenic and anti-proliferative effects in CTB cells. Human extravillous CTB cells of the line Sw-71, derived from first trimester chorionic villus tissue, were incubated with 0, 0.1, 1, 10, and 100 nM of each of three CTSs (MBG, cinobufatalin (CINO) and ouabain (OUB)) for 48 h. Thereafter, levels of pro-angiogenic (vascular endothelial growth factor (VEGF165), placental growth factor (PlGF)) and anti-angiogenic (soluble fms-like tyrosine kinase-1 (sFlt-1), soluble endoglin (sEng)) factors were measured in culture media using ELISA kits. Expression of three receptors (VEGF receptor 1 (VEGFR1), angiogenic angiotensin type 1 receptor (AT1) and anti-angiogenic angiotensin type 2 receptor (AT2)) were assayed using immunoblotting (western blots) in cell lysates. sFlt-1 and sEng secretion were increased while VEGF165 and PIGF were decreased in the culture media of CTB cells treated with 1 nM or more of each CTSs (p < 0.01 for each). The AT2 receptor expression was up-regulated (p < 0.05) in CTB cells treated with 1 nM or more of MBG and CINO and with 100 nM OUB, while AT1 and VEGFR1 expressions decreased (p < 0.05) with 1 nM or more of MBG and 10 nM or more of CINO and OUB. CTSs influence extravillous CTB cells to induce an anti-angiogenic and anti-proliferative profile.
Product Description
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Glutamine