MilliporeSigma
  • Quantifying the biomechanics of conception: L-selectin-mediated blastocyst implantation mechanics with engineered "trophospheres".
Quantifying the biomechanics of conception: L-selectin-mediated blastocyst implantation mechanics with engineered "trophospheres".
Tissue engineering. Part A (2013-08-10)
Robert W Yucha, Monika Jost, Dianne Rothstein, Noreen Robertson, Michele S Marcolongo
ABSTRACT
An estimated 12% of women in the United States suffer from some form of infertility. In vitro fertilization (IVF) is the most common treatment for infertility encompassing over 99% of all assisted reproductive technologies. However, IVF has a low success rate. Live birth rates using IVF can range from 40% in women younger than 35 years to 4% in women older than 42 years. Costs for a successful IVF outcome can be upward of $61,000. The low success rate of IVF has been attributed to the inability of the blastocyst to implant to the uterus. Blastocyst implantation is initiated by L-selectin expressing cells, trophoblasts, binding to L-selectin ligands, primarily sialyl Lewis X (sLeX), on the uterine surface endometrium. Legal and ethical considerations have limited the research on human subjects and tissues, whereas animal models are costly or do not properly mimic human implantation biochemistry. In this work, we describe a cellular model system for quantifying L-selectin adhesion mechanics. L-selectin expression was confirmed in Jeg-3, JAR, and BeWo cell lines, with only Jeg-3 cells exhibiting surface expression. Jeg-3 cells were cultured into three-dimensional spheres, termed "trophospheres," as a mimic to human blastocysts. Detachment assays using a custom-built parallel plate flow chamber show that trophospheres detach from sLeX functionalized slides with 2.75 × 10(-3) dyn of force and 7.5 × 10(-5) dyn-cm of torque. This work marks the first time a three-dimensional cell model has been utilized for quantifying L-selectin binding mechanics related to blastocyst implantation.
Product Description
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
β-Estradiol, ≥98%