MilliporeSigma
  • The soluble receptor for vitamin B12 uptake (sCD320) increases during pregnancy and occurs in higher concentration in urine than in serum.
The soluble receptor for vitamin B12 uptake (sCD320) increases during pregnancy and occurs in higher concentration in urine than in serum.
PloS one (2013-09-10)
Omar Abuyaman, Birgitte H Andreasen, Camilla Kronborg, Erik Vittinghus, Ebba Nexo
ABSTRACT
Cellular uptake of vitamin B12 (B12) demands binding of the vitamin to transcobalamin (TC) and recognition of TC-B12 (holoTC) by the receptor CD320, a receptor expressed in high quantities on human placenta. We have identified a soluble form of CD320 (sCD320) in serum and here we present data on the occurrence of this soluble receptor in both serum and urine during pregnancy. We examined serum from twenty-seven pregnant women (cohort 1) at gestational weeks 13, 24 and 36 and serum and urine samples from forty pregnant women (cohort 2) tested up to 8 times during gestational weeks 17-41. sCD320, holoTC, total TC and complex formation between holoTC and sCD320 were measured by in-house ELISA methods, while creatinine was measured on the automatic platform Cobas 6000. Size exclusion chromatography was performed on a Superdex 200 column. Median (range) of serum sCD320 increased from 125 (87-839) pmol/L (week 15) to reach a peak value of 199 (72-672) pmol/L (week 35) then dropped back to its baseline level just before birth (week 40). Around one third of sCD320 was precipitated with holoTC at all-time points studied. The urinary concentration of sCD320 was around two fold higher than in serum. Urinary sCD320/creatinine ratio correlated with serum sCD320 and reached a peak median level of 53 (30-101) pmol/mmol creatinine (week 35). sCD320 present in serum and urine showed the same elution pattern upon size exclusion chromatography. We report for the first time that sCD320 is present in urine and in a higher concentration than in serum and that serum and urine sCD320 increase during pregnancy. The high urinary concentration and the strong correlation between urinary and serum sCD320 suggests that sCD320 is filtered in the kidney.
Product Description
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard