MilliporeSigma
Rab3a promotes brain tumor initiation and progression.
Molecular biology reports (2014-06-27)
Jun-Kyum Kim, Seung-Yup Lee, Chang-Won Park, Suk-Hwang Park, Jinlong Yin, Jaebong Kim, Jae-Bong Park, Jae-Yong Lee, Hyunggee Kim, Sung-Chan Kim
ABSTRACT
The Rab protein family is composed of small GTP-binding proteins involved in intracellular vesicle trafficking. In particular, Rab3a which is one of four Rab3 proteins (a, b, c, and d isoforms) is associated with synaptic vesicle trafficking in normal brain. However, despite the elevated level of Rab3a in tumors, its role remains unclear. Here we report a tumorigenic role of Rab3a in brain tumors. Elevated level of Rab3a expression in human was confirmed in both glioma cell lines and glioblastoma multiforme patient specimens. Ectopic Rab3a expression in glioma cell lines and primary astrocytes promoted cell proliferation by increasing cyclin D1 expression, induced resistance to anti-cancer drug and irradiation, and accelerated foci formation in soft agar and tumor formation in nude mice. The overexpression of Rab3a augmented the tumorsphere-forming ability of glioma cells and p53(-/-) astrocytes and increased expression levels of various stem cell markers. Taken together, our results indicate that Rab3a is a novel oncogene involved in glioma initiation and progression.
Sigma-Aldrich
MISSION® esiRNA, targeting human CCND1
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
Staurosporine, VETRANAL®, analytical standard
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)