MilliporeSigma
  • Regulation of Hfq mRNA and protein levels in Escherichia coli and Pseudomonas aeruginosa by the Burkholderia cenocepacia MtvR sRNA.
Regulation of Hfq mRNA and protein levels in Escherichia coli and Pseudomonas aeruginosa by the Burkholderia cenocepacia MtvR sRNA.
PloS one (2014-06-06)
Christian G Ramos, André M Grilo, Sílvia A Sousa, Joana R Feliciano, Paulo J P da Costa, Jorge H Leitão
ABSTRACT
Small non-coding RNAs (sRNAs) are important players of gene expression regulation in bacterial pathogens. MtvR is a 136-nucleotide long sRNA previously identified in the human pathogen Burkholderia cenocepacia J2315 and with homologues restricted to bacteria of the Burkholderia cepacia complex. In this work we have investigated the effects of expressing MtvR in Escherichia coli and Pseudomonas aeruginosa. Results are presented showing that MtvR negatively regulates the hfq mRNA levels in both bacterial species. In the case of E. coli, this negative regulation is shown to involve binding of MtvR to the 5'-UTR region of the hfqEc mRNA. Results presented also show that expression of MtvR in E. coli and P. aeruginosa originates multiple phenotypes, including reduced resistance to selected stresses, biofilm formation ability, and increased susceptibility to various antibiotics.
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Trimethoprim, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥98.5%