MilliporeSigma
  • Expression of a novel peptide derived from PCNA damages DNA and reverses cisplatin resistance.
Expression of a novel peptide derived from PCNA damages DNA and reverses cisplatin resistance.
Cancer chemotherapy and pharmacology (2014-09-06)
Robert G Lingeman, Robert J Hickey, Linda H Malkas
ABSTRACT
An 8 amino acid peptide sequence derived from proliferating cell nuclear antigen (PCNA) has been shown to effectively kill several breast cancer and neuroblastoma cell lines when added exogenously to cell cultures. In this study, the expression of the 8 amino acid peptide sequence (caPeptide) was placed under control of a tetracycline responsive promoter in MDA-MB-231 cells. Endogenous expression of the peptide resulted in an increase in genomic DNA damage. CaPeptide induction combined with treatment of sublethal doses of cisplatin resulted in a marked increase in death of the cisplatin-resistant MDA-MB-231 cell line. CaPeptide was found to interact with POLD3, one of the subunits of DNA polymerase delta necessary for binding to PCNA. These results suggest an important line of inquiry into the possible role that caPeptide might play in the reversal of cisplatin resistance in breast and other cancers. This is of particular interest in those cancers where cisplatin is the first line of chemotherapy and where the acquisition of resistance is a common malady.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
SAFC
L-Glutamine