MilliporeSigma
  • Comparative analysis of gene expression in normal and degenerative human tendon cells: effects of cyclic strain.
Comparative analysis of gene expression in normal and degenerative human tendon cells: effects of cyclic strain.
Foot & ankle international (2014-06-25)
Woo Jin Choi, Min Sung Park, Kwang Hwan Park, Jean-Paul Courneya, Jin Sun Cho, Lew C Schon, Jin Woo Lee
ABSTRACT
Tendinopathy is a clinical problem for which treatment shows mixed results and treatment options are limited. Gene expression signatures early in the mechanotransduction pathway can accurately predict risk and correlate with different clinical outcomes. Studies aimed at elucidating the molecular mechanisms of tendinopathy have focused on small cohorts of genes that show an incomplete picture of the degeneration process. This study compared the effect of cyclic strain on gene expression in tendon cells from normal tendon and chronically painful areas of tendinopathy in 3 patients. We measured a panel of mechanotransduction genes and cytoskeletal tensional balance with and without cyclic strain, which disrupts connective tissue synthetic-degradative equilibrium. Normal and degenerative tendons were obtained from patients undergoing surgery to treat chronic painful tendinopathy. A cyclic strain model was established to measure cytoskeletal tensional homeostasis. Prior to cyclic strain, the normal tendon cells exhibited varying patterns of elevated expression of 7 genes compared with degenerative tendon cells. In response to cyclic strain, gene expression of COL1A1, ITGA6, CTNNA1, and CLEC3B was up-regulated in normal tendon cells. Cyclic strain had no effect on degenerative tendon cells. Cyclic strain exacerbated the inhibition of protein synthesis in both cell types, especially in the degenerative tendon cells. Alterations in the pattern of gene expression are suggestive of a dynamic equilibrium between synthesis and degradation, whereby cell adhesion molecules are predominantly up-regulated to facilitate cellular reorientation in response to their altered functional environment. These data might have future applications, including the identification of markers for early diagnosis, targets for drug design, and indicators for treatment responsiveness and prognosis.
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water