MilliporeSigma
  • YB-1 regulates Sox2 to coordinately sustain stemness and tumorigenic properties in a phenotypically distinct subset of breast cancer cells.
YB-1 regulates Sox2 to coordinately sustain stemness and tumorigenic properties in a phenotypically distinct subset of breast cancer cells.
BMC cancer (2014-06-03)
Karen Jung, Fang Wu, Peng Wang, Xiaoxia Ye, Bassam S Abdulkarim, Raymond Lai
ABSTRACT
Sox2, a transcription factor and an embryonic stem cell marker, has been implicated in the pathogenesis of breast cancer (BC). YB-1 is another transcription factor that has been shown to promote stemness in BC cells. Western blotting, quantitative PCR, and siRNAs were used to query the regulatory relationships between YB-1, Sox2, and their downstream targets. Chromatin immunoprecipitation was used to detect YB-1 interactions at the Sox2 promoter. Mammosphere and soft agar assays were used to assess the phenotypic consequences of YB-1 knockdown. Here, we report that YB-1 regulates Sox2. YB-1 was found to bind to the SOX2 promoter and down-regulate its expression in MCF7 and ZR751. The regulatory interaction between YB-1 and Sox2 was drastically different between the two phenotypically distinct cell subsets, purified based on their differential response to a Sox2 reporter. They are referred to as the reporter unresponsive (RU) cells and the reporter responsive (RR) cells. Upon siRNA knockdown of YB-1, RU cells showed an increase in Sox2 expression but no change in Sox2 reporter activity; in contrast, RR cells exhibited increased expression and reporter activity of Sox2. Correlating with these findings, YB-1 knockdown induced a differential response in the expression of genes known to be regulated by both Sox2 and YB-1 (e.g. CCND1 and ITGA6). For instance, in response to YB-1 knockdown, CCND1 and ITGA6 expression were decreased or unchanged in RU cells but paradoxically increased in RR cells. Compared to RU cells, RR cells were significantly more resistant to the suppression of mammosphere formation due to YB-1 knockdown. Importantly, mammospheres derived from parental MCF7 cells treated with YB-1 siRNA knockdown exhibited higher expression levels of SOX2 and its downstream targets. To conclude, in a subset of BC cells, namely RR cells, YB-1 regulates Sox2 to coordinately maintain stemness and tumorigenic properties.
Sigma-Aldrich
Anti-SOX2 antibody produced in rabbit
Sigma-Aldrich
Anti-sox2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
GSK-3 Inhibitor XVI, GSK-3 Inhibitor XVI - CAS 252917-06-9, is a cell-permeable, potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC₅₀ = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively).
Sigma-Aldrich
Anti-SOX2 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Insulin-like Growth Factor-I human, IGF-I, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
RSK Inhibitor, SL0101, The RSK Inhibitor, SL0101 controls the biological activity of RSK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)