MilliporeSigma
  • Liver X Receptor activation delays chondrocyte hypertrophy during endochondral bone growth.
Liver X Receptor activation delays chondrocyte hypertrophy during endochondral bone growth.
Osteoarthritis and cartilage (2014-05-24)
M M-G Sun, F Beier
ABSTRACT
Activation of the Liver X Receptor (LXR) has recently been identified as a therapeutic strategy for osteoarthritis (OA). Human OA articular cartilage explants show decreased LXR expression, and LXRβ-null mice display OA-like symptoms. LXR agonist administration to OA articular cartilage explants suppresses proteoglycan degradation and restores LXR-activated transcription. We aimed to investigate the effect of LXR activation on chondrocyte differentiation to elucidate the molecular mechanisms behind its protection against OA. The specific LXR agonist, GW3965, was used to examine the effect of LXR activation on chondrocyte differentiation. Tibia organ cultures were used to examine the effect of LXR activation on bone growth and growth plate morphology, followed by immunohistochemical analysis. In ATDC5 and micromass cultures, chondrocyte differentiation was examined through cellular staining and proliferation assays. Various chondrogenic markers were analyzed by real-time reverse-transcription polymerase chain reaction (qRT-PCR) in micromass RNA. Chondrocyte hypertrophy was suppressed by GW3965 treatment, as shown by decreased hypertrophic zone length in the tibial growth plate, decreased alkaline phosphatase staining in ATDC5 and micromass cultures, and down regulation of Col10a1, Mmp13 and Runx2 expression. Increased proliferation in treated ATDC5 cells and up-regulation of Col2a1 expression in treated micromass cultures suggest hypertrophy is suppressed secondary to prolonged proliferation. Decreased p57 levels in treated growth plates suggest this to be due to cell-cycle exit delay. Our findings regarding LXR's role in cartilage development provide insight into how LXR activation prevents cartilage breakdown, further solidifying its potential as a therapeutic target of OA.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Selenium, pellets, < 5mm, ≥99.999%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Supelco
N,N-Dimethylformamide, analytical standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%