MilliporeSigma
  • Prostaglandin E-prostanoid4 receptor mediates angiotensin II-induced (pro)renin receptor expression in the rat renal medulla.
Prostaglandin E-prostanoid4 receptor mediates angiotensin II-induced (pro)renin receptor expression in the rat renal medulla.
Hypertension (Dallas, Tex. : 1979) (2014-05-29)
Fei Wang, Xiaohan Lu, Kexin Peng, Yaomin Du, Shu-Feng Zhou, Aihua Zhang, Tianxin Yang
ABSTRACT
Angiotensin II (Ang II) stimulates (pro)renin receptor (PRR) expression in the renal collecting duct, triggering the local renin response in the distal nephron. Our recent study provided evidence for involvement of cyclooxygenase-2-prostaglandin E2 pathway in Ang II-dependent stimulation of PRR expression in the collecting duct. Here, we tested the role of E-prostanoid (EP) subtypes acting downstream of cyclooxygenase-2 in this phenomenon. In primary rat inner medullary collecting duct cells, Ang II treatment for 12 hours induced a 1.8-fold increase in the full-length PRR protein expression. To assess the contribution of EP receptor, the cell was pretreated with specific EP receptor antagonists: SC-51382 (for EP1), L-798106 (for EP3), L-161982 (for EP4), and ONO-AE3-208 (ONO, a structurally distinct EP4 antagonist). The upregulation of PRR expression by Ang II was consistently abolished by L-161982 and ONO and partially suppressed by SC-51382 but was unaffected by L-798106. The PRR expression was also significantly elevated by the EP4 agonist CAY10598 in the absence of Ang II. Sprague-Dawley rats were subsequently infused for 1 or 2 weeks with vehicle, Ang II alone, or in combination with ONO. Ang II infusion induced parallel increases in renal medullary PRR protein and renal medullary and urinary renin activity and total renin content, all of which were blunted by ONO. Both tail cuff plethysmography and telemetry demonstrated attenuation of Ang II hypertension by ONO. Overall, these results have established a crucial role of the EP4 receptor in mediating the upregulation of renal medullary PRR expression and renin activity during Ang II hypertension.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
L-798106, ≥98% (HPLC)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Renin Substrate Tetradecapeptide porcine, ≥97% (HPLC)