MilliporeSigma
  • Anoctamin 1 calcium-activated chloride channel downregulates estrogen production in mouse ovarian granulosa cells.
Anoctamin 1 calcium-activated chloride channel downregulates estrogen production in mouse ovarian granulosa cells.
Endocrinology (2014-05-16)
Meiyan Sun, Yujie Sui, Lihua Li, Weiheng Su, Feng Hao, Qi Zhu, Weihua Di, Hongwen Gao, Tonghui Ma
ABSTRACT
Calcium-dependent chloride conductances have been described in chicken and human granulosa cells (GCs) and may be involved in steroidogenesis. However, the molecular identities of corresponding chloride channels remain unknown. The purpose of this study was to explore the expression and function of the Anoctamin 1 (ANO1) calcium-activated chloride channel (CaCC) in mouse ovary. ANO1 mRNA and protein expression was identified in mouse ovary GCs by RT-PCR, immunoblot, and immunostaining. Patch-clamp analysis on freshly isolated GCs identified an outwardly rectifying Ca(2+)-activated Cl(-) current that was completely blocked by a selective ANO1 inhibitor T16Ainh-A01. Knockdown of ANO1 mRNA or incubation with a selective inhibitor T16Ainh-A01 enhanced estradiol production, whereas a selective ANO1 activator Eact significantly inhibited estradiol production in primary cultured GCs. The ANO1 expression or activation increases the phosphorylation of ERK1/2 and decreases aromatase expression. The ANO1 expression level is remarkably higher at the proestrous and estrous stages in the estrous cycle. In vivo study indicated a profound induction of ANO1 expression in ovarian GCs by pregnant mare serum gonadotropin (PMSG) that can be further augmented by hCG treatment, suggesting that both FSH and LH may upregulate ANO1 expression at the proestrous and estrous stages. ANO1 expression was remarkably reduced in DHEA-induced PCOS ovary. These data identified for the first time the expression of ANO1 Ca(2+) activated Cl(-) channel in mouse ovarian GCs and determined its negative regulation on estrogen production possibly through MEK-ERK signaling cascade. The present study provided new insights into the molecular mechanisms for the regulation of folliculogenesis and ovulation.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Selenium, pellets, < 5mm, ≥99.999%
SAFC
Sodium chloride solution, 5 M
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
ITS Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Estradiol, meets USP testing specifications
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material