MilliporeSigma
  • Exposure-dose-response of Tellina deltoidalis to contaminated estuarine sediments 3. Selenium spiked sediments.
Exposure-dose-response of Tellina deltoidalis to contaminated estuarine sediments 3. Selenium spiked sediments.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2014-07-11)
Anne M Taylor, William A Maher
ABSTRACT
The metalloid selenium is an essential element which at slightly elevated concentrations is toxic and mutagenic. In Australia the burning of coal for power generation releases selenium into estuarine environments where it accumulates in sediments. The relationship between selenium exposure, dose and response was investigated in the deposit feeding, benthic, marine bivalve Tellina deltoidalis. Bivalves were exposed in microcosms for 28 days to individual selenium spiked sediments, 0, 5 and 20 μg/g dry mass. T. deltoidalis accumulated selenium from spiked sediment but not in proportion to the sediment selenium concentrations. The majority of recovered subcellular selenium was associated with the nuclei and cellular debris fraction, probably as protein bound selenium associated with plasma and selenium bound directly to cell walls. Selenium exposed organisms had increased biologically detoxified selenium burdens which were associated with both granule and metallothionein like protein fractions, indicating selenium detoxification. Half of the biologically active selenium was associated with the mitochondrial fraction with up to 4 fold increases in selenium in exposed organisms. Selenium exposed T. deltoidalis had significantly reduced GSH:GSSG ratios indicating a build-up of oxidised glutathione. Total antioxidant capacity of selenium exposed T. deltoidalis was significantly reduced which corresponded with increased lipid peroxidation, lysosomal destabilisation and micronuclei frequency. Clear exposure-dose-response relationships have been demonstrated for T. deltoidalis exposed to selenium spiked sediments, supporting its suitability for use in selenium toxicity tests using sub-lethal endpoints.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Acid phosphatase Assay Kit, 1 kit sufficient for 1,000 assays (multiwell plates), 1 kit sufficient for 100 assays (tubes)
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC