MilliporeSigma
  • Dysregulation of system xc(-) expression induced by mutant huntingtin in a striatal neuronal cell line and in R6/2 mice.
Dysregulation of system xc(-) expression induced by mutant huntingtin in a striatal neuronal cell line and in R6/2 mice.
Neurochemistry international (2014-07-09)
Natalie M Frederick, Julie Bertho, Kishan K Patel, Geraldine T Petr, Ekaterina Bakradze, Sylvia B Smith, Paul A Rosenberg
ABSTRACT
Oxidative stress has been implicated in the pathogenesis of Huntington's disease (HD), however, the origin of the oxidative stress is unknown. System xc(-) plays a role in the import of cystine to synthesize the antioxidant glutathione. We found in the STHdh(Q7/Q7) and STHdh(Q111/Q111) striatal cell lines, derived from neuronal precursor cells isolated from knock-in mice containing 7 or 111 CAG repeats in the huntingtin gene, that there is a decrease in system xc(-) function. System xc(-) is composed of two proteins, the substrate specific transporter, xCT, and an anchoring protein, CD98. The decrease in function in system xc(-) that we observed is caused by a decrease in xCT mRNA and protein expression in the STHdh(Q111/Q111) cells. In addition, we found a decrease in protein and mRNA expression in the transgenic R6/2 HD mouse model at 6weeks of age. STHdh(Q111/Q111) cells have lower basal levels of GSH and higher basal levels of ROS. Acute inhibition of system xc(-) causes greater increase in oxidative stress in the STHdh(Q111/Q111) cells than in the STHdh(Q7/Q7) cells. These results suggest that a defect in the regulation of xCT may be involved in the pathogenesis of HD by compromising xCT expression and increasing susceptibility to oxidative stress.
Product Description
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid