MilliporeSigma
  • Alterations of the RRAS and ERCC1 genes at 19q13 in gemistocytic astrocytomas.
Alterations of the RRAS and ERCC1 genes at 19q13 in gemistocytic astrocytomas.
Journal of neuropathology and experimental neurology (2014-09-06)
Takashi Ohta, Young-Ho Kim, Ji-Eun Oh, Kaishi Satomi, Naosuke Nonoguchi, Kathy Keyvani, Daniela Pierscianek, Ulrich Sure, Michel Mittelbronn, Werner Paulus, Anne Vital, Hideaki Yokoo, Kerrie McDonald, Paul Kleihues, Nicolas Nazaret, Fabienne Barbet, Joel Lachuer, Hiroko Ohgaki
ABSTRACT
Gemistocytic astrocytoma (World Health Organization grade II) is a rare variant of diffuse astrocytoma that is characterized by the presence of neoplastic gemistocytes and has a significantly less favorable prognosis. Other than frequent TP53 mutations (>80%), little is known about its molecular profile. Here, we show that gemistocytic astrocytomas carry a lower frequency of IDH mutations than fibrillary astrocytomas (74% vs 92%; p = 0.0255) but have profiles similar to those of fibrillary astrocytomas with respect to TERT promoter mutations (5% vs 0%), 1p/19q loss (10% vs 8%), and loss of heterozygosity 10q (10% vs 12%). Exome sequencing in 5 gemistocytic astrocytomas revealed homozygous deletion of genes at 19q13 (i.e. RRAS [related RAS viral oncogene homolog; 2 cases] and ERCC1 [excision repair cross-complementing rodent repair deficiency, complementation group 1; 1 case]). Further screening showed RRAS homozygous deletion in 7 of 42 (17%) gemistocytic astrocytomas and in 3 of 24 (13%) IDH1 mutated secondary glioblastomas. Patients with gemistocytic astrocytoma and secondary glioblastoma with an RRAS deletion tended to have shorter survival rates than those without deletion. Differential polymerase chain reaction and methylation-specific polymerase chain reaction revealed an ERCC1 homozygous deletion or promoter methylation in 10 of 42 (24%) gemistocytic astrocytomas and in 8 of 24 (33%) secondary glioblastomas. Alterations in RRAS and ERCC1 appear to be typical in gemistocytic astrocytomas and secondary glioblastomas, since they were not present in 49 fibrillary astrocytomas or 30 primary glioblastomas.
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, ≥99% (HPLC)
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%