MilliporeSigma
  • Microfluidic genotyping by rapid serial PCR and high-speed melting analysis.
Microfluidic genotyping by rapid serial PCR and high-speed melting analysis.
Clinical chemistry (2014-08-15)
Scott O Sundberg, Carl T Wittwer, Renée M Howell, Jarkko Huuskonen, Robert J Pryor, Jared S Farrar, Heather M Stiles, Robert A Palais, Ivor T Knight
ABSTRACT
Clinical molecular testing typically batches samples to minimize costs or uses multiplex lab-on-a-chip disposables to analyze a few targets. In genetics, multiple variants need to be analyzed, and different work flows that rapidly analyze multiple loci in a few targets are attractive. We used a microfluidic platform tailored to rapid serial PCR and high-speed melting (HSM) to genotype 4 single nucleotide variants. A contiguous stream of master mix with sample DNA was pulsed with each primer pair for serial PCR and melting. Two study sites each analyzed 100 samples for F2 (c.*97G>A), F5 (c.1601G>A), and MTHFR (c.665C>T and c.1286A>C) after blinding for genotype and genotype proportions. Internal temperature controls improved melting curve precision. The platform's liquid-handling system automated PCR and HSM. PCR and HSM were completed in a total of 12.5 min. Melting was performed at 0.5 °C/s. As expected, homozygous variants were separated by melting temperature, and heterozygotes were identified by curve shape. All samples were correctly genotyped by the instrument. Follow-up testing was required on 1.38% of the assays for a definitive genotype. We demonstrate genotyping accuracy on a novel microfluidic platform with rapid serial PCR and HSM. The platform targets short turnaround times for multiple genetic variants in up to 8 samples. It is also designed to allow automatic and immediate reflexive or repeat testing depending on results from the streaming DNA. Rapid serial PCR provides a flexible genetic work flow and is nicely matched to HSM analysis.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Betaine monohydrate, ≥99.0% (NT)
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Betaine monohydrate, ≥99%
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female