MilliporeSigma
  • Induction of human CYP3A4 by huperzine A, ligustrazine and oridonin through pregnane X receptor-mediated pathways.
Induction of human CYP3A4 by huperzine A, ligustrazine and oridonin through pregnane X receptor-mediated pathways.
Die Pharmazie (2014-07-31)
Yi-Wen Zhang, Mei-Hua Bao, Guo Wang, Qiang Qu, Hong-Hao Zhou
ABSTRACT
The pregnane X receptor (PXR) is a key regulator of CYP3A4, which is involved in catalyzing the metabolic conversion of a number of endogenous substrates. In this study, we screened 22 compounds isolated from traditional Chinese herbal medicines using luciferase reporter gene assays for inspecting their capabilities in inducing PXR-mediated transactivation of CYP3A4 expression. In addition, the mRNA and protein expressions of CYP3A4 and PXR as well as the enzymatic activites of CYP3A4 were analyzed by real-time PCR, Western blot analysis and UPLC-MS/MS-based metabolite assay in LS174T cells. Huperzine A, ligustrazine and oridonin were identified to be the inducers of CYP3A4. These compounds induced the CYP3A4 reporter luciferase activity, and up-regulated CYP3A4 mRNA and protein levels significantly. Besides, huperzine A, ligustrazine and oridonin significantly up-regulated enzymatic activities of CYP3A4. However, the three compounds showed no effects on PXR mRNA and protein expression. To our knowledge, it is the first identification of these three compounds as PXR activators to induce CYP3A4. These results indicate that huperzine A, ligustrazine and oridonin induced CYP3A4 expression and activation via PXR dependent pathways, and might contribute to drug-drug interactions.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Oridonin, ≥98% (HPLC), solid
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Supelco
2,3,5,6-Tetramethylpyrazine, analytical standard
Rifampicin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
2,3,5,6-Tetramethylpyrazine, 98%
Sigma-Aldrich
2,3,5,6-Tetramethylpyrazine, ≥98%, FG
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
2,3,5,6-Tetramethylpyrazine, natural, ≥98%, FG