MilliporeSigma
  • Serum proteomic analysis reveals potential serum biomarkers for occupational medicamentosa-like dermatitis caused by trichloroethylene.
Serum proteomic analysis reveals potential serum biomarkers for occupational medicamentosa-like dermatitis caused by trichloroethylene.
Toxicology letters (2014-06-25)
Peiwu Huang, Xiaohu Ren, Zhijun Huang, Xifei Yang, Wenxu Hong, Yanfang Zhang, Hang Zhang, Wei Liu, Haiyan Huang, Xinfeng Huang, Desheng Wu, Linqing Yang, Haiyan Tang, Li Zhou, Xuan Li, Jianjun Liu
ABSTRACT
Trichloroethylene (TCE) is an industrial solvent with widespread occupational exposure and also a major environmental contaminant. Occupational medicamentosa-like dermatitis induced by trichloroethylene (OMLDT) is an autoimmune disease and it has become one major hazard in China. In this study, sera from 3 healthy controls and 3 OMLDT patients at different disease stages were used for a screening study by 2D-DIGE and MALDI-TOF-MS/MS. Eight proteins including transthyretin (TTR), retinol binding protein 4 (RBP4), haptoglobin, clusterin, serum amyloid A protein (SAA), apolipoprotein A-I, apolipoprotein C-III and apolipoprotein C-II were found to be significantly altered among the healthy, acute-stage, healing-stage and healed-stage groups. Specifically, the altered expression of TTR, RBP4 and haptoglobin were further validated by Western blot analysis and ELISA. Our data not only suggested that TTR, RBP4 and haptoglobin could serve as potential serum biomarkers of OMLDT, but also indicated that measurement of TTR, RBP4 and haptoglobin or their combination could help aid in the diagnosis, monitoring the progression and therapy of the disease.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Trichloroethylene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Trichloroethylene, analytical standard, stabilized with 30 – 50 ppm Diisopropylamine
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%
Sigma-Aldrich
Trichloroethylene, ACS reagent, ≥99.5%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Trichloroethylene, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%