MilliporeSigma
  • Avocado consumption enhances human postprandial provitamin A absorption and conversion from a novel high-β-carotene tomato sauce and from carrots.
Avocado consumption enhances human postprandial provitamin A absorption and conversion from a novel high-β-carotene tomato sauce and from carrots.
The Journal of nutrition (2014-06-06)
Rachel E Kopec, Jessica L Cooperstone, Ralf M Schweiggert, Gregory S Young, Earl H Harrison, David M Francis, Steven K Clinton, Steven J Schwartz
ABSTRACT
Dietary lipids have been shown to increase bioavailability of provitamin A carotenoids from a single meal, but the effects of dietary lipids on conversion to vitamin A during absorption are essentially unknown. Based on previous animal studies, we hypothesized that the consumption of provitamin A carotenoids with dietary lipid would enhance conversion to vitamin A during absorption compared with the consumption of provitamin A carotenoids alone. Two separate sets of 12 healthy men and women were recruited for 2 randomized, 2-way crossover studies. One meal was served with fresh avocado (Persea americana Mill), cultivated variety Hass (delivering 23 g of lipid), and a second meal was served without avocado. In study 1, the source of provitamin A carotenoids was a tomato sauce made from a novel, high-β-carotene variety of tomatoes (delivering 33.7 mg of β-carotene). In study 2, the source of provitamin A carotenoids was raw carrots (delivering 27.3 mg of β-carotene and 18.7 mg of α-carotene). Postprandial blood samples were taken over 12 h, and provitamin A carotenoids and vitamin A were quantified in triglyceride-rich lipoprotein fractions to determine baseline-corrected area under the concentration-vs.-time curve. Consumption of lipid-rich avocado enhanced the absorption of β-carotene from study 1 by 2.4-fold (P < 0.0001). In study 2, the absorption of β-carotene and α-carotene increased by 6.6- and 4.8-fold, respectively (P < 0.0001 for both). Most notably, consumption of avocado enhanced the efficiency of conversion to vitamin A (as measured by retinyl esters) by 4.6-fold in study 1 (P < 0.0001) and 12.6-fold in study 2 (P = 0.0013). These observations highlight the importance of provitamin A carotenoid consumption with a lipid-rich food such as avocado for maximum absorption and conversion to vitamin A, especially in populations in which vitamin A deficiency is prevalent. This trial was registered at clinicaltrials.gov as NCT01432210.
Product Description
Supelco
β-Carotene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Carotene, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
β-Carotene, synthetic, ≥93% (UV), powder
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Supelco
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, reference material, Cerilliant®
Supelco
α-Carotene, analytical standard