MilliporeSigma
  • Studies on the mechanism of testicular dysfunction in the early stage of a streptozotocin induced diabetic rat model.
Studies on the mechanism of testicular dysfunction in the early stage of a streptozotocin induced diabetic rat model.
Biochemical and biophysical research communications (2014-05-31)
Yongde Xu, Hongen Lei, Ruili Guan, Zhezhu Gao, Huixi Li, Lin Wang, Weidong Song, Bing Gao, Zhongcheng Xin
ABSTRACT
Streptozotocin (STZ) induced diabetic model has been widely used to study the effects of diabetes mellitus (DM) on male infertility, but it remains unclear whether the responses in this model are due to hyperglycemia or STZ per se. This study was designed to investigate the mechanism of STZ on testicular dysfunction. In the present study, sperm characteristics, serum testosterone, steroidogenic enzymes (StAR and 3β-HSD), and the vimentin apical extension of sertoli cells decreased significantly in the STZ group compared with those in the normal controls (p<0.05), while Johnsen's score, testicular lipid peroxidation, spermatogenic cell apoptosis, and the expressions of NF-κB and Wnt4 significantly increased (p<0.05). Insulin replacement mainly restored the decreased serum testosterone and steroidogenic enzymes, but not other parameters. The results indicated that spermatogenic dysfunction in the early stage of STZ-induced diabetic rats was due to direct STZ cytotoxicity to sertoli cells, which could be regulated by Wnt4 and NF-κB, while steroidogenic dysfunction might be a direct or indirect consequence of insulin deficiency. The results suggested that STZ-induced diabetic model, at least in the early stage, is not suitable to study the diabetes-related spermatogenic dysfunction.
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-PCNA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®