MilliporeSigma
  • Use of topical liposomes containing meglumine antimoniate (Glucantime) for the treatment of L. major lesion in BALB/c mice.
Use of topical liposomes containing meglumine antimoniate (Glucantime) for the treatment of L. major lesion in BALB/c mice.
Experimental parasitology (2014-05-02)
S A Moosavian Kalat, A Khamesipour, N Bavarsad, M Fallah, Z Khashayarmanesh, E Feizi, K Neghabi, A Abbasi, M R Jaafari
ABSTRACT
Treatment of cutaneous leishmaniasis (CL) is a dream for the patients, health care authorities and scientists. The aim of this study was to develop a topical liposomal meglumine antimoniate (MA, Glucantime™) (Lip-MA) formulation and evaluate the therapeutic effects of the preparation on lesion induced by Leishmania major in BALB/c mice. Liposomes containing 22.5% MA (6.4% Sb(+5)) with and without oleic acid (LMA-OA and LMA) were formulated using fusion method plus homogenization and characterized for the size and encapsulation efficiency. The penetration of MA from the LMA-OA and LMA formulations through and into the skin was checked in vitro using Franz diffusion cells fitted with mouse skin at 37°C for 8h. The in vitro permeation data showed that almost 1.5% of formulations applied in the mouse skin were penetrated and the amount retained in the skin was about 65%. The 50% effective dose of LMA and LMA-OA against amastigotes of L. major was 46.36 and 41.01 μg/ml, respectively. LMA or LMA-OA was used topically twice a day for 4 weeks to treat the lesion induced by L. major in susceptible BALB/c mice. The results showed a significantly (P<0.001) smaller lesion size in the treated groups of mice compared to the control groups which received either empty liposomes or phosphate-buffered saline (PBS). The spleen parasite burden was significantly (P<0.001) lower in the treated groups compared to the control groups receiving either empty liposomes or PBS at the end of the treatment period. However, when the treatment was stopped, the lesion size progressed and spleen parasite burden increased in LMA and LMA-OA groups, but still was significantly less than the control groups (P<0.05). There was no significant difference between the two formulations of LMA and LMA-OA. The results suggested that topical liposomes containing MA might be an appropriate choice for clinical trials for the treatment of CL.
Product Description
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
SAFC
HEPES
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(±)-α-Tocopherol, analytical standard
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Meglumine, 99.0-100.5% dry basis, meets USP testing specifications
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material