MilliporeSigma
  • Attenuation of natural killer cell functions by capsaicin through a direct and TRPV1-independent mechanism.
Attenuation of natural killer cell functions by capsaicin through a direct and TRPV1-independent mechanism.
Carcinogenesis (2014-04-20)
Hun Sik Kim, Hyung-Joon Kwon, Gye Eun Kim, Mi-Hyang Cho, Seung-Yong Yoon, Alexander J Davies, Seog Bae Oh, Heuiran Lee, Young Keol Cho, Chul Hyun Joo, Seog Woon Kwon, Sun Chang Kim, Yoo Kyum Kim
ABSTRACT
The assessment of the biological activity of capsaicin, the compound responsible for the spicy flavor of chili pepper, produced controversial results, showing either carcinogenicity or cancer prevention. The innate immune system plays a pivotal role in cancer pathology and prevention; yet, the effect of capsaicin on natural killer (NK) cells, which function in cancer surveillance, is unclear. This study found that capsaicin inhibited NK cell-mediated cytotoxicity and cytokine production (interferon-γ and tumor necrosis factor-α). Capsaicin impaired the cytotoxicity of NK cells, thereby inhibiting lysis of standard target cells and gastric cancer cells by modulating calcium mobilization in NK cells. Capsaicin also induced apoptosis in gastric cancer cells, but that effect required higher concentrations and longer exposure times than those required to trigger NK cell dysfunction. Furthermore, capsaicin inhibited the cytotoxicity of isolated NK cells and of an NK cell line, suggesting a direct effect on NK cells. Antagonists of transient receptor potential vanilloid subfamily member 1 (TRPV1), a cognate capsaicin receptor, or deficiency in TRPV1 expression failed to prevent the defects induced by capsaicin in NK cells expressing functional TRPV1. Thus, the mechanism of action of capsaicin on NK cells is largely independent of TRPV1. Taken together, capsaicin may have chemotherapeutic potential but may impair NK cell function, which plays a central role in tumor surveillance.
Product Description
Supelco
Capsaicin, analytical standard
Supelco
Capsaicin, Pharmaceutical Secondary Standard; Certified Reference Material
Capsaicin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC), Molecular Biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Capsazepine, ≥98% (HPLC), solid
Sigma-Aldrich
Propidium iodide solution