MilliporeSigma
  • Polymorphism in the ADRB2 gene explains a small portion of intersubject variability in pain relative to cervical dilation in the first stage of labor.
Polymorphism in the ADRB2 gene explains a small portion of intersubject variability in pain relative to cervical dilation in the first stage of labor.
Anesthesiology (2014-04-10)
Abdullah S Terkawi, William M Jackson, Shehnaz Hansoti, Rabeena Tabassum, Pamela Flood
ABSTRACT
Variability in labor pain has been associated with demographic, clinical, and psychological factors. Polymorphisms of the β2-adrenergic receptor gene (ADRB2) influence sensitivity to experimental pain in humans and are a risk factor for chronic pain. The authors hypothesized that polymorphisms in ADRB2 may influence labor pain. After Institutional Review Board approval and written informed consent, the authors prospectively obtained hourly pain reports from 233 nulliparous parturients during the first stage of labor, of which 199 were included in the current analysis. DNA from blood samples was genotyped at polymorphisms in the genes for the β2-adrenergic receptor, the μ opioid receptor subtype 1, catechol-O-methyltransferase, fatty acid amide hydrolase, and the oxytocin receptor. Labor pain as a function of cervical dilation was modeled with previously described methods. Patient covariates, ADRB2 genotype, and obstetrical and anesthesia treatment were evaluated as covariates in the model. Labor pain more rapidly became severe in parturients heterozygous or homozygous for the G allele at rs1042714 in the ADRB2 gene. Labor pain increased more rapidly after artificial rupture of membranes, augmentation with oxytocin, and in younger women. Inclusion of covariates explained approximately 10% of the variability between subjects. ADRB2 genotype explained less than 1% of the intersubject variability. ADRB2 genotype correlates with labor pain but explained less than 1% of the intersubject variance in the model.
Product Description
Oxytocin, European Pharmacopoeia (EP) Reference Standard
USP
Oxytocin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Oxytocin acetate salt hydrate, ≥97% (HPLC)