MilliporeSigma
  • Protein expression profiling during wallerian degeneration after rat sciatic nerve injury.
Protein expression profiling during wallerian degeneration after rat sciatic nerve injury.
Muscle & nerve (2013-10-15)
Meiyuan Li, Pingan Zhang, Weimin Guo, Huaiqin Li, Xiaosong Gu, Dengbing Yao
ABSTRACT
Wallerian degeneration (WD) is an important area of research in modern neuroscience. Many protein expressions are regulated by differentially expressed genes in WD, but the precise mechanisms are elusive. In this study, we profiled differentially expressed proteins in WD after rat sciatic nerve injury using an antibody array. Functional analysis positively identified cell proliferation, regulation of cell proliferation, and immune system processes. Kyoto Encyclopedia of Genes and Genomes (KEGG) analysis revealed molecular networks related mainly to cytokine-cytokine receptor interaction, the mitogen-activated proteinkinase (MAPK) signaling pathway, apoptosis, the toll-like receptor (TLR) signaling pathway, and the Janus kinase (Jak) - signal transducer and activator of transcription (STAT) signaling pathway. Interactions between these differential proteins were well established and regulated by the key factors transforming growth factor beta 1 (TGF-β1), toll-like receptor 4 (TLR4), Fas ligand (FasL), and 5'-AMP-activated protein kinase catalytic subunit alpha-1 (PRKAA1). These results provide information related to functional analysis of differentially expressed genes during WD.
Product Description
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Biotin, European Pharmacopoeia (EP) Reference Standard
SAFC
BIOTIN
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Biotin, ≥99.0% (T)
USP
Biotin, United States Pharmacopeia (USP) Reference Standard