MilliporeSigma
  • Flavoprotein structure and mechanism. 4. Xanthine oxidase and xanthine dehydrogenase.
Flavoprotein structure and mechanism. 4. Xanthine oxidase and xanthine dehydrogenase.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (1995-08-01)
R Hille, T Nishino
ABSTRACT
Xanthine oxidase and xanthine dehydrogenase are enzymes involved in the metabolism of purines and pyrimidines in various organisms. Their relationship to one another has been the subject of considerable debate, primarily because of their proposed roles in ischemia/reperfusion damage in tissues. Differences in the kinetics and oxidation-reduction behavior of the two forms are accounted for by the presence in the dehydrogenase of a binding site for NAD+, as well as a substantially lower reduction potential for the flavin FADH./FADH2 couple of the dehydrogenase relative to the oxidase. This review presents recent advances of our understanding of the biochemistry and molecular biology of these systems, including a model for the overall morphology of xanthine oxidizing enzymes. The evidence that the two enzymes represent alternate forms of the same gene product, in some cases reversibly interconvertible between one another, is discussed.
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Sigma-Aldrich
Xanthine, ≥99%