MilliporeSigma
  • Increasing plasma [K+] by intravenous potassium infusion reduces NCC phosphorylation and drives kaliuresis and natriuresis.
Increasing plasma [K+] by intravenous potassium infusion reduces NCC phosphorylation and drives kaliuresis and natriuresis.
American journal of physiology. Renal physiology (2014-03-07)
Srinivas Rengarajan, Donna H Lee, Young Taek Oh, Eric Delpire, Jang H Youn, Alicia A McDonough
ABSTRACT
Dietary potassium loading results in rapid kaliuresis, natriuresis, and diuresis associated with reduced phosphorylation (p) of the distal tubule Na(+)-Cl(-) cotransporter (NCC). Decreased NCC-p inhibits NCC-mediated Na(+) reabsorption and shifts Na(+) downstream for reabsorption by epithelial Na(+) channels (ENaC), which can drive K(+) secretion. Whether the signal is initiated by ingesting potassium or a rise in plasma K(+) concentration ([K(+)]) is not understood. We tested the hypothesis, in male rats, that an increase in plasma [K(+)] is sufficient to reduce NCC-p and drive kaliuresis. After an overnight fast, a single 3-h 2% potassium (2%K) containing meal increased plasma [K(+)] from 4.0 ± 0.1 to 5.2 ± 0.2 mM; increased urinary K(+), Na(+), and volume excretion; decreased NCC-p by 60%; and marginally reduced cortical Na(+)-K(+)-2Cl(-) cotransporter (NKCC) phosphorylation 25% (P = 0.055). When plasma [K(+)] was increased by tail vein infusion of KCl to 5.5 ± 0.1 mM over 3 h, significant kaliuresis and natriuresis ensued, NCC-p decreased by 60%, and STE20/SPS1-related proline alanine-rich kinase (SPAK) phosphorylation was marginally reduced 35% (P = 0.052). The following were unchanged at 3 h by either the potassium-rich meal or KCl infusion: Na(+)/H(+) exchanger 3 (NHE3), NHE3-p, NKCC, ENaC subunits, and renal outer medullary K(+) channel. In summary, raising plasma [K(+)] by intravenous infusion to a level equivalent to that observed after a single potassium-rich meal triggers renal kaliuretic and natriuretic responses, independent of K(+) ingestion, likely driven by decreased NCC-p and activity sufficient to shift sodium reabsorption downstream to where Na(+) reabsorption and flow drive K(+) secretion.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sigma-Aldrich
Fructose-6-phosphate Kinase from Bacillus stearothermophilus, Type VII, lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)