MilliporeSigma
Survival of patients with transformed lymphoma in the rituximab era.
Annals of hematology (2014-01-15)
Hany R Guirguis, Matthew C Cheung, Eugenia Piliotis, David Spaner, Neil L Berinstein, Kevin Imrie, Liying Zhang, Rena Buckstein
ABSTRACT
In the pre-rituximab era, transformation of indolent B-cell lymphoma to diffuse large B-cell lymphoma (DLBCL) was associated with an extremely poor outcome and a median post-transformation survival ranging from 1 to 2 years. We evaluated the impact of rituximab-cyclophosphamide, adriamycin, vincristine, prednisone (R-CHOP) on the survival outcomes of transformed lymphoma compared with de novo DLBCL. Between 2002 and 2010, 317 DLBCL patients who were consecutively diagnosed and treated with R-CHOP were identified at our institution. Patients with transformed lymphoma were included if they had not previously received R-CHOP. Patient characteristics, treatment, and outcome data were retrospectively collected. Sixty patients (19 %) had transformed lymphoma of which 37 (62 %) had transformed from follicular lymphoma, 50 (83 %) were chemotherapy naïve, and 58 (96 %) were rituximab naïve at the time of treatment. With a median follow-up of 31.4 months, 231 patients achieved either complete response or complete response unconfirmed (73 %) with no significant difference between de novo DLBCL (n = 192, 75 %) and the transformed group (n = 39, 65 %) (P = 0.25). Six patients (15 %) relapsed in the transformed group at a median time to relapse of 29.3 months. The 2-year and 5-year overall survivals for all patients were 82 and 72 %, respectively. The overall and progression-free survivals for transformed lymphoma and de novo DLBCL were not statistically different (P = 0.45 and P = 0.38, respectively). With R-CHOP chemotherapy, the prognosis of transformed lymphoma in patients with minimal chemotherapy exposure for indolent disease is similar to that of de novo DLBCL.
Product Description
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals