MilliporeSigma
Myocardial Mycn is essential for mouse ventricular wall morphogenesis.
Developmental biology (2012-10-16)
Cristina Harmelink, Yin Peng, Paige DeBenedittis, Hanying Chen, Weinian Shou, Kai Jiao
ABSTRACT
MYCN is a highly conserved transcription factor with multifaceted roles in development and disease. Mutations in MYCN are associated with Feingold syndrome, a developmental disorder characterized in part by congenital heart defects. Mouse models have helped elucidate MYCN functions; however its cardiac-specific roles during development remain unclear. We employed a Cre/loxp strategy to uncover the specific activities of MYCN in the developing mouse myocardium. Myocardial deletion of Mycn resulted in a thin-myocardial wall defect with dramatically reduced trabeculation. The mutant heart defects strongly resemble the phenotype caused by disruption of BMP10 and Neuregulin-1 (NRG1) signaling pathways, two central mediators of myocardial wall development. Our further examination showed that expression of MYCN is regulated by both BMP and NRG1 signaling. The thin-wall defect in mutant hearts is caused by a reduction in both cell proliferation and cell size. MYCN promotes cardiomyocyte proliferation through regulating expression of cell cycle regulators (including CCND1, CCND2, and ID2) and promotes cardiomyocyte growth through regulating expression of p70S6K. In addition, expression of multiple sarcomere proteins is altered in Mycn myocardial-inactivation embryos, indicating its essential role for proper cardiomyocyte differentiation. In summary, Mycn acts downstream of BMP and NRG1 cardiogenic signaling pathways to promote normal myocardial wall morphogenesis.
Sigma-Aldrich
AG 1478, A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.
Sigma-Aldrich
Anti-Actin, Cardiac antibody, Mouse monoclonal, clone AC1-20.4.2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid