MilliporeSigma
  • Matrix metalloproteinase (MMP)-1 induces lung alveolar epithelial cell migration and proliferation, protects from apoptosis, and represses mitochondrial oxygen consumption.
Matrix metalloproteinase (MMP)-1 induces lung alveolar epithelial cell migration and proliferation, protects from apoptosis, and represses mitochondrial oxygen consumption.
The Journal of biological chemistry (2013-08-02)
Iliana Herrera, José Cisneros, Mariel Maldonado, Remedios Ramírez, Blanca Ortiz-Quintero, Elena Anso, Navdeep S Chandel, Moisés Selman, Annie Pardo
ABSTRACT
Idiopathic pulmonary fibrosis is a devastating lung disorder of unknown etiology. Although its pathogenesis is unclear, considerable evidence supports an important role of aberrantly activated alveolar epithelial cells (AECs), which produce a large variety of mediators, including several matrix metalloproteases (MMPs), which participate in fibroblast activation and lung remodeling. MMP-1 has been shown to be highly expressed in AECs from idiopathic pulmonary fibrosis lungs although its role is unknown. In this study, we explored the role of MMP-1 in several AECs functions. Mouse lung epithelial cells (MLE12) transfected with human Mmp-1 showed significantly increased cell growth and proliferation at 36 and 48 h of culture (p < 0.01). Also, MMP-1 promoted MLE12 cell migration through collagen I, accelerated wound closing, and protected cells from staurosporine- and bleomycin-induced apoptosis compared with mock cells (p < 0.01). MLE12 cells expressing human MMP-1 showed a significant repression of oxygen consumption ratio compared with the cells with the empty vector. As under hypoxic conditions hypoxia-inducible factor-1α (HIF-1α) mediates a transition from oxidative to glycolytic metabolism, we analyzed activation of HIF-1α. Ηigher activation of this factor was detected in MMP-1-transfected cells under normoxia and hypoxia. Likewise, a significant decrease of both total and mitochondrial reactive oxygen species was observed in MMP-1-transfected cells. Paralleling these findings, attenuation of MMP-1 expression by shRNA in A549 (human) AECs markedly reduced proliferation and migration (p < 0.01) and increased the oxygen consumption ratio. These findings indicate that epithelial expression of MMP-1 inhibits mitochondrial function, increases HIF-1α expression, decreases reactive oxygen species production, and contributes to a proliferative, migratory, and anti-apoptotic AEC phenotype.
Product Description
Supelco
Staurosporine, VETRANAL®, analytical standard
Sigma-Aldrich
Anti-MMP-1 (Ab-1) Mouse mAb (41-1E5), liquid, clone 41-1E5, Calbiochem®
Sigma-Aldrich
Staurosporine from Streptomyces sp., ≥98% (HPLC), film
Sigma-Aldrich
Staurosporine from Streptomyces sp., ≥95% (HPLC)