MilliporeSigma
  • Interplay between Homeobox proteins and Polycomb repressive complexes in p16INK⁴a regulation.
Interplay between Homeobox proteins and Polycomb repressive complexes in p16INK⁴a regulation.
The EMBO journal (2013-03-05)
Nadine Martin, Nikolay Popov, Francesca Aguilo, Ana O'Loghlen, Selina Raguz, Ambrosius P Snijders, Gopuraja Dharmalingam, Side Li, Efstathia Thymiakou, Thomas Carroll, Bernd B Zeisig, Chi Wai Eric So, Gordon Peters, Vasso Episkopou, Martin J Walsh, Jesús Gil
ABSTRACT
The INK4/ARF locus regulates senescence and is frequently altered in cancer. In normal cells, the INK4/ARF locus is found silenced by Polycomb repressive complexes (PRCs). Which are the mechanisms responsible for the recruitment of PRCs to INK4/ARF and their other target genes remains unclear. In a genetic screen for transcription factors regulating senescence, we identified the homeodomain-containing protein HLX1 (H2.0-like homeobox 1). Expression of HLX1 extends cellular lifespan and blunts oncogene-induced senescence. Using quantitative proteomics, we identified p16(INK4a) as the key target mediating the effects of HLX1 in senescence. HLX1 represses p16(INK4a) transcription by recruiting PRCs and HDAC1. This mechanism has broader implications, as HLX1 also regulates a subset of PRC targets besides p16(INK4a). Finally, sampling members of the Homeobox family, we identified multiple genes with ability to repress p16(INK4a). Among them, we found HOXA9 (Homeobox A9), a putative oncogene in leukaemia, which also recruits PRCs and HDAC1 to regulate p16(INK4a). Our results reveal an unexpected and conserved interplay between homeodomain-containing proteins and PRCs with implications in senescence, development and cancer.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys9) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-p21WAF1/Cip1antibody, Mouse monoclonal, clone CP74, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)