MilliporeSigma
  • 2,6-Dimethyloctanedioic acid--a metabolite of phytanic acid in Refsum's disease.
2,6-Dimethyloctanedioic acid--a metabolite of phytanic acid in Refsum's disease.
Clinical chemistry (1983-03-01)
J Greter, S Lindstedt, G Steen
ABSTRACT
The urine of two patients with Refsum's disease consistently contained 2,6-dimethyloctanedioic acid, a compound not normally found in human urine. In addition, their urines contained increased concentrations of 3-methylhexanedioic acid. These two compounds may be formed from phytanic acid by an initial omega-oxidation and subsequent beta-oxidations. It was calculated that this oxidation pathway may metabolize at least 30 mg of phytanic acid per day.
Sigma-Aldrich
3-Methyladipic acid, 99%
Sigma-Aldrich
Methylsuccinic acid, 99%
Sigma-Aldrich
Pipecolinic acid, 98%
Sigma-Aldrich
Phytanic acid, ≥96%, mixture of isomers