MilliporeSigma
  • Development and significance of cysteamine and propionitrile models of duodenal ulcer.
Development and significance of cysteamine and propionitrile models of duodenal ulcer.
Chronobiology international (1987-01-01)
S Szabo, G Pihan
ABSTRACT
Cysteamine is widely used in rodents to induce duodenal ulcer. Herein, the pathogenesis of duodenal ulceration in its earliest stages was reviewed using findings from cysteamine- and propionitrile-induced duodenal ulcer in rodent models, especially taking into account changes in the secretion of gastric acid, duodenal and pancreatic bicarbonate as well as gastroduodenal motility. The effect of cysteamine-HCl in inducing ulcers in rats is circadian rhythm-dependent. The effect is greatest from just before the end of diurnal rest to just after the start of nocturnal activity. The chronobiologic effect may be in part due to the circadian rhythm-dependent increased gastric acid production from cysteamine. Titratable acidity was found to be twice as great in the gastric juice of rodents when cysteamine was given by injection at 2000 (just after the start of nocturnal activity) in comparison to when given at 0800 or 1200 (at the beginning or middle span of daily rest). Further studies have shown that adrenalectomy of rats 7 days before cysteamine administration obliterated the observed circadian susceptibility to ulcer formation. Duodenal ulceration, at least in the cysteamine model, appears to be under chronobiologic neuroendocrine control or influence, seemingly mediated by the adrenal glands.
Sigma-Aldrich
Cysteamine hydrochloride, BioXtra
Supelco
Propionitrile, analytical standard
Sigma-Aldrich
Cysteamine, ≥98.0% (RT)
Sigma-Aldrich
Cysteamine, ~95%
Sigma-Aldrich
Propionitrile, 99%
Sigma-Aldrich
Propionitrile, purum, ≥99.0% (GC)