MilliporeSigma
  • The risks associated with aprotinin use: a retrospective study of cardiac cases in Nova Scotia.
The risks associated with aprotinin use: a retrospective study of cardiac cases in Nova Scotia.
Canadian journal of anaesthesia = Journal canadien d'anesthesie (2012-11-08)
Robert E G Riddell, Karen J Buth, John A Sullivan
ABSTRACT
In light of the concerns about the safety of aprotinin, we wanted to determine if aprotinin use during cardiac surgery was associated with an increased risk of mortality and morbidity compared with the use of tranexamic acid (TXA). We hypothesized that use of aprotinin is associated with a higher risk of adverse outcomes than use of TXA in our patient population. In this retrospective study at a single surgical centre, we examined primary in-hospital outcomes of postoperative mortality, new acute renal failure, and perioperative blood transfusion, and we also investigated secondary outcomes of stroke, infection, and prolonged stay in the intensive care unit (ICU). The effect of the type of antifibrinolytic on outcome was evaluated for aprotinin cases matched 1:1 with TXA cases using propensity score. This study included 3,340 patients who received antifibrinolytics during cardiac surgery (376 patients received aprotinin and 2,964 patients received TXA). Patients who received aprotinin were more often elderly and female; they were more commonly presented with congestive heart failure, atrial fibrillation, renal failure, and lower hemoglobin, and they underwent complex and/or urgent surgery. In the matched sample, in-hospital mortality was significantly higher in the aprotinin group (10.9%) compared with the TXA group (5.9%), and ICU stay >72 hr was significantly increased in the aprotinin group (30.0%) compared with the TXA group (21.7%). There was no significant difference in blood product administration between the two groups. Aprotinin was associated with an increased risk of in-hospital mortality and morbidity following cardiac surgery, and aprotinin was not associated with a decrease in blood product requirements. Continued use of aprotinin in cardiac surgery should follow careful consideration, weighing the demonstrated risks and potential advantages compared with other TXA.
Sigma-Aldrich
Aprotinin from bovine lung, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE)
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-7 TIU/mg solid
Sigma-Aldrich
Aprotinin from bovine lung
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg