MilliporeSigma
  • Transmetallation between metal-only Lewis pairs: a new rhodium alane complex.
Transmetallation between metal-only Lewis pairs: a new rhodium alane complex.
Chemical communications (Cambridge, England) (2012-09-20)
Jürgen Bauer, Holger Braunschweig, Krzysztof Radacki
ABSTRACT
In this communication, synthesis of a rhodium alane Lewis adduct, [Cp(Me(3)P)(2)Rh→AlCl(3)], is reported. Given that direct synthesis using aluminium trichloride failed, a convenient transmetallation reaction was applied. The new MOLP was analysed by single crystal X-ray diffraction, multinuclear NMR spectroscopy and density functional theory calculations.
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, sublimed, ≥98%
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum chloride, reagent grade, 98%
Sigma-Aldrich
Aluminum chloride solution, 1.0 M in nitrobenzene
Sigma-Aldrich
Aluminum chloride hydrate, 99.999% trace metals basis