MilliporeSigma
  • Nano-sized manganese oxide: a proposed catalyst for water oxidation in the reaction of some manganese complexes and cerium(IV) ammonium nitrate.
Nano-sized manganese oxide: a proposed catalyst for water oxidation in the reaction of some manganese complexes and cerium(IV) ammonium nitrate.
Dalton transactions (Cambridge, England : 2003) (2012-07-19)
Mohammad Mahdi Najafpour, Atefeh Nemati Moghaddam
ABSTRACT
According to UV-visible spectroscopy, X-ray diffraction spectrometry, dynamic light scattering, Fourier transform infrared spectroscopy, electron paramagnetic resonance spectroscopy, transmission electron microscopy, scanning electron microscopy and X-ray photoelectron spectroscopy, nano-sized manganese oxides are proposed as active catalysts for water oxidation in the reaction of some manganese complexes and cerium(IV) ammonium nitrate.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%