MilliporeSigma
  • Ryanodine receptor current amplitude controls Ca2+ sparks in cardiac muscle.
Ryanodine receptor current amplitude controls Ca2+ sparks in cardiac muscle.
Circulation research (2012-05-26)
Tao Guo, Dirk Gillespie, Michael Fill
ABSTRACT
In cardiac muscle, Ca(2+)-induced Ca(2+) release (CICR) from the sarcoplasmic reticulum (SR) is mediated by ryanodine receptor (RyR) Ca(2+) release channels. The inherent positive feedback of CICR is normally well-controlled. Understanding this control mechanism is a priority because its malfunction has life-threatening consequences. We show that CICR local control is governed by SR Ca(2+) load, largely because load determines the single RyR current amplitude that drives inter-RyR CICR. We differentially manipulated single RyR Ca(2+) flux amplitude and SR Ca(2+) load in permeabilized ventricular myocytes as an endogenous cell biology model of the heart. Large RyR-permeable organic cations were used to interfere with Ca(2+) conductance through the open RyR pore. Single-channel studies show this attenuates current amplitude without altering other aspects of RyR function. In cells, the same experimental maneuver increased resting SR Ca(2+) load. Despite the increased load, Ca(2+) spark (inter-RyR CICR events) frequency decreased and sparks terminated earlier. Spark local control follows single RyR current amplitude, not simply SR Ca(2+) load. Spark frequency increases with load because spontaneous RyR openings at high loads produce larger currents (ie, a larger CICR trigger signal). Sparks terminate when load falls to the point at which single RyR current amplitude is no longer sufficient to sustain inter-RyR CICR. Thus, RyRs that spontaneously close no longer reopen and local Ca(2+) release ends.
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine hydrochloride, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine hydrochloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%