MilliporeSigma
  • A simple, reproducible and sensitive spectrophotometric method to estimate microalgal lipids.
A simple, reproducible and sensitive spectrophotometric method to estimate microalgal lipids.
Analytica chimica acta (2012-04-10)
Yimin Chen, Seetharaman Vaidyanathan
ABSTRACT
Quantification of total lipids is a necessity for any study of lipid production by microalgae, especially given the current interest in microalgal carbon capture and biofuels. In this study, we employed a simple yet sensitive method to indirectly measure the lipids in microalgae by measuring the fatty acids (FA) after saponification. The fatty acids were reacted with triethanolamine-copper salts (TEA-Cu) and the ternary TEA-Cu-FA complex was detected at 260 nm using a UV-visible spectrometer without any colour developer. The results showed that this method could be used to analyse low levels of lipids in the range of nano-moles from as little as 1 mL of microalgal culture. Furthermore, the structure of the TEA-Cu-FA complex and related reaction process are proposed to better understand this assay. There is no special instrument required and the method is very reproducible. To the best of our knowledge, this is the first report of the use of UV absorbance of copper salts with FA as a method to estimate lipids in algal cultures. It will pave the way for a more convenient assay of lipids in microalgae and can readily be expanded for estimating lipids in other biological systems.
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine hydrochloride, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine hydrochloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%