MilliporeSigma
  • Conversion of hexose into 5-hydroxymethylfurfural in imidazolium ionic liquids with and without a catalyst.
Conversion of hexose into 5-hydroxymethylfurfural in imidazolium ionic liquids with and without a catalyst.
Carbohydrate research (2011-04-05)
Quan Cao, Xingcui Guo, Shengxi Yao, Jing Guan, Xiaoyan Wang, Xindong Mu, Dongke Zhang
ABSTRACT
Conversion of fructose and glucose into 5-hydroxymethylfurfural (HMF) was investigated in various imidazolium ionic liquids, including 1-butyl-3-methylimidazolium chloride (BmimCl), 1-hexyl-3-methylimidazolium chloride (HmimCl), 1-octyl-3-methylimidazolium chloride (OmimCl), 1-benzyl-3-methylimidazolium chloride (BemimCl), 1-Butyl-2,3-dimethylimidazolium chloride (BdmimCl), and 1-butyl-3-methylimidazolium p-toluenesulfonate (BmimPS). The acidic C-2 hydrogen of imidazolium cations was shown to play a major role in the dehydration of fructose in the absence of a catalyst, such as sulfuric acid or CrCl(3). Both the alkyl groups of imidazolium cations and the type of anions affected the reactivity of the carbohydrates. Although, except BmimCl and BemimCl, other four ionic liquids could only achieve not more than 25% HMF yields without an additional catalyst, 60-80% HMF yields were achieved in HmimCl, BdmimCl, and BmimPS in the presence of sulfuric acid or CrCl(3) in sufficient quantities.
Sigma-Aldrich
Chromium(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
1-Butyl-2,3-dimethylimidazolium chloride, ≥97.0% (HPLC/AT)
Sigma-Aldrich
Chromium(II) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Chromium(II) chloride, 95%