MilliporeSigma
  • Immobilization of horseradish peroxidase on nanoporous copper and its potential applications.
Immobilization of horseradish peroxidase on nanoporous copper and its potential applications.
Bioresource technology (2010-08-17)
Huajun Qiu, Lu Lu, Xirong Huang, Zhonghua Zhang, Yinbo Qu
ABSTRACT
Nanoporous copper (NPC) with a pore size of 100-200 nm was prepared by simply dealloying Al(60)Cu(40) alloy in a 5 wt.% HCl solution. The NPC was characterized by scanning electron microscopy and nitrogen adsorption techniques. Horseradish peroxidase (HRP) was immobilized on NPC by adsorption. Compared with free enzyme, the thermal stability of the immobilized enzyme was greatly improved due to the multiple attachments between the enzyme molecule and the NPC surface. After 2h incubation at 50 degrees C, the immobilized HRP retained ca. 90% of the initial activity while only ca. 10% initial activity remained for the free enzyme. The interaction between HRP and the porous surface also made the K(m) and K(cat) values of the immobilized enzyme increase (from 0.43 to 0.80 mM) and decrease (from 8.1 x 10(3) to 2.2 x 10(3)min(-1)), respectively. Based on the good electric conductivity and electrocatalytic activity of the NPC electrode, an electrochemical biosensor for O-phenylenediamine (OPD) was made. The calibration curve of the biosensor was linear from 0.5 microM to 14.5 microM OPD with a sensitivity of 0.37 microA microM(-1). The stability and reproducibility of the biosensor were also demonstrated to be good. When positioned at -0.45 V for 200 s, its current response toward 10 microM OPD remained ca. 80% of its initial value. For five HRP-loaded NPC electrodes, the relative standard deviation (RSD) of the current response toward 10 microM OPD was ca. 4.5%. All these results indicated that NPC was a good support for the HRP immobilization and its low price would facilitate its large-scale application.
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%