MilliporeSigma
  • Facile assembly of hybrid materials containing polyoxometalate cluster anions and organic dye cations: crystal structures and initial spectral characterization.
Facile assembly of hybrid materials containing polyoxometalate cluster anions and organic dye cations: crystal structures and initial spectral characterization.
Chemical communications (Cambridge, England) (2008-01-23)
Jingli Xie, Brendan F Abrahams, Anthony G Wedd
ABSTRACT
Salts of the pararosaniline dye cation and four polyoxometalate cluster anions have been isolated under both ambient and hydrothermal conditions; structural and initial spectroscopic data are consistent with significant perturbation of ion electronic states induced by charge-assisted N-H---O hydrogen bonds.
Sigma-Aldrich
Basic Fuchsin, Dye content >85 %
Sigma-Aldrich
Basic Fuchsin, certified by the BSC, Dye content ≥88 %
Sigma-Aldrich
Pararosaniline acetate, Dye content 90 %
Sigma-Aldrich
Pararosaniline hydrochloride, >85.0% (HPLC)