MilliporeSigma
  • Joint stress of chlorimuron-ethyl and cadmium on wheat Triticum aestivum at biochemical levels.
Joint stress of chlorimuron-ethyl and cadmium on wheat Triticum aestivum at biochemical levels.
Environmental pollution (Barking, Essex : 1987) (2006-03-15)
Mei-E Wang, Qi-Xing Zhou
ABSTRACT
Biochemical responses to joint stress of chlorimuron-ethyl and cadmium (Cd) in wheat Triticum aestivum were examined. The joint action of chlorimuron-ethyl and Cd weakened the inhibition of Cd or chlorimuron-ethyl on the formation of chlorophyll. It was deduced that wheat plants had the capability to protect themselves by increasing the activity of the antioxidant enzyme peroxidase (POD) with the exposure time. The joint effect of chlorimuron-ethyl and Cd on the superoxide dismutase (SOD) activity in leaves was additive, while the joint effect on the SOD activity in roots was determined by the interaction of chlorimuron-ethyl and Cd in wheat. It was also concluded that the change of malondialdehyde (MDA) content in wheat might not be a good biomarker in the oxidative damage by chlorimuron-ethyl, while a decrease in the soluble protein content and POD activity in roots could be considered as a biomarker in the damage of wheat by chlorimuron-ethyl and Cd.
Product Description
Supelco
Chlorimuron ethyl, PESTANAL®, analytical standard
Supelco
Chlorimuron ethyl, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland