MilliporeSigma
  • An improved CTAB-ammonium acetate method for total RNA isolation from cotton.
An improved CTAB-ammonium acetate method for total RNA isolation from cotton.
Phytochemical analysis : PCA (2012-05-04)
Lu Zhao, Qi Ding, Jun Zeng, Fu-Rong Wang, Jun Zhang, Shou-Jin Fan, Xin-Qiang He
ABSTRACT
Cotton is an important economic crop. Genetic, developmental and molecular studies of cotton require high-quality total RNA from different tissues. Due to the richness in polyphenols and polysaccharides, the Trizol-based methods and other commercial kits are unsuitable for RNA isolation from cotton. Available methods are generally laborious and time-consuming. To develop an easy, simple and rapid cetyltrimethylammonium bromide (CTAB)-ammonium acetate protocol that takes less time and obtains high yield and quality of RNA from polysaccharide- and polyphenol-rich cotton tissues. Based on the original CTAB protocol, we used phenol-chloroform and chloroform-isoamyl alcohol to remove proteins, polysaccharides and polyphenols, and ammonium acetate to precipitate RNA, reducing the incubation time prior to RNA precipitation. After adding ammonium acetate to precipitate RNA, all centrifugation steps (14000 × g) were carried out at 4°C to avoid degradation. The procedure took only 1.5 h and was suitable for different cotton tissues. The A(260) : A(280) ratios ranged from 1.80 to 1.85 with clear 28 s and 18 s ribosomal RNA bands in 1.2% agarose gel. The isolated RNA was usable for downstream molecular studies, such as reverse transcription polymerase chain reaction (PCR) and real-time quantitative PCR. The CTAB-ammonium acetate method is easy, rapid, low-cost and effective for high-quality RNA isolation from polysaccharide- and polyphenol-rich cotton tissues.
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate solution, 7.5 M
Sigma-Aldrich
Chloroform – isoamyl alcohol mixture, BioUltra, 49:1, ≥99.5% (chloroform + isoamyl alcohol, GC)