MilliporeSigma
  • Notch inhibits expression of the Krüppel-like factor 4 tumor suppressor in the intestinal epithelium.
Notch inhibits expression of the Krüppel-like factor 4 tumor suppressor in the intestinal epithelium.
Molecular cancer research : MCR (2008-12-17)
Amr M Ghaleb, Gaurav Aggarwal, Agnieszka B Bialkowska, Mandayam O Nandan, Vincent W Yang
ABSTRACT
The zinc finger-containing transcription factor, Krüppel-like factor 4 (KLF4), inhibits cell proliferation. An in vivo tumor-suppressive role for KLF4 is shown by the recent finding that Klf4 haploinsufficiency in Apc(Min/+) mice promotes intestinal tumorigenesis. Studies also show that KLF4 is required for the terminal differentiation of goblet cells in the mouse intestine. The Notch signaling pathway suppresses goblet cell formation and is up-regulated in intestinal tumors. Here, we investigated the relationship between Notch signaling and KLF4 expression in intestinal epithelial cells. The rate of proliferation of HT29 human colon cancer cells was reduced when treated with the gamma-secretase inhibitor dibenzazepine to inhibit Notch signaling or small interfering RNA directed against Notch. KLF4 levels were increased in dibenzazepine-treated or Notch small interfering RNA-treated cells. Conversely, overexpression of Notch in HT29 cells reduced KLF4 levels, suppressed KLF4 promoter activity, and increased proliferation rate. Treatment of Apc(Min/+) mice with dibenzazepine resulted in a 50% reduction in the number of intestinal adenomas compared with the vehicle-treated group (P < 0.001). Both the normal-appearing intestinal mucosa and adenomas obtained from dibenzazepine-treated Apc(Min/+) mice had increased goblet cell numbers and Klf4 staining accompanied by reduced cyclin D1 and Ki-67 staining when compared with those from vehicle-treated mice. Results of these studies indicate that Notch signaling suppresses KLF4 expression in intestinal tumors and colorectal cancer cells. Inhibition of Notch signaling increases KLF4 expression and goblet cell differentiation and reduces proliferation and tumor formation. KLF4 is therefore a potential mediator for the antitumor effect of Notch inhibitors such as dibenzazepine.
Sigma-Aldrich
Alcian Blue 8GX, powder
Sigma-Aldrich
Alcian Blue 8GX
Sigma-Aldrich
Alcian Blue 8GX, BioReagent, suitable for detection of glycoproteins
Sigma-Aldrich
Deshydroxy LY-411575, ≥98% (HPLC)
Sigma-Aldrich
Alcian Blue 8GX, certified by the BSC